Merck
Hepatotoxic effects of mycotoxin combinations in mice.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-12-03)
Lv-Hui Sun, Ming-yan Lei, Ni-Ya Zhang, Ling Zhao, Christopher Steven Krumm, De-Sheng Qi
ZUSAMMENFASSUNG
This study was performed to assess the individual and combined toxic effects of aflatoxin B1 (AFB1), zearalenone (ZEA) and deoxynivalenol (DON) within the liver of mice. A total of 56 4-week-old weanling female mice were divided into seven groups (n = 8). For 2 weeks, each group received an oral administration of either solvent (control), AFB1, ZEA, DON, AFB1 + ZEA, AFB1 + DON or ZEA + DON per day. The results showed that AFB1, ZEA and DON induced liver injury, indicated by elevated relative liver weight, activities of alanine aminotransferase (ALT) and/or aspartate aminotransferase (AST), as well as decreased albumin (ALB) and/or total protein (TP) concentration in the serum. These mycotoxins also decreased hepatic total antioxidant capacity (T-AOC), and/or increased the concentration of malondialdehyde (MDA). Moreover, AFB1 + DON displayed synergistic effects, while AFB1 + ZEA displayed antagonistic effects on those parameters previously described. Furthermore, the apoptotic potential was demonstrated associated with an upregulation of the apoptotic genes Caspase-3 and Bax, along with a downregulation of the antiapoptotic gene Bcl-2 in liver. In conclusion, this study provides a better understanding of the toxic effects of AFB1, ZEA, DON, alone or in combinations on the liver of mice, which could contribute to the risk assessment of these mycotoxins in food and feed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Aflatoxin B1, from Aspergillus flavus
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4,4′-Diaminodiphenylmethan, ≥97.0% (GC)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Ethanol
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Dimethylsulfoxid, analytical standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard