Merck
  • Cross-desensitization of CCR1, but not CCR2, following activation of the formyl peptide receptor FPR1.
Cross-desensitization of CCR1, but not CCR2, following activation of the formyl peptide receptor FPR1.
Journal of immunology (Baltimore, Md. : 1950) (2014-04-30)
Filip Bednar, Changcheng Song, Giuseppe Bardi, William Cornwell, Thomas J Rogers
ZUSAMMENFASSUNG
The cross-regulation of G protein-coupled receptors (GPCRs) plays an important role in the immune response. Studies from several laboratories have suggested that a hierarchy of sensitivities to cross-desensitization exists for the chemoattractant GPCRs. We carried out experiments to study the capacity of the formyl peptide receptor-1 (FPR1) to desensitize chemokine receptors CCR1 and CCR2. Our results show that activation of FPR1 resulted in the desensitization and partial internalization of CCR1, but not CCR2, in both primary human monocytes and HEK293 cells coexpressing CCR1, CCR2, and FPR1 (HR1R2F cells). The desensitization of CCR1 by FPR1 stimulation was not due to the simple depletion of the Ca(2+) stores, but was dependent on activation of protein kinase C. Furthermore, we found that the cross-desensitization of CCR1 by FPR1 was associated with CCR1 phosphorylation and moderate reduction of CCR1 cell-surface expression. In contrast, CCR2 was not phosphorylated or internalized after FPR1 activation. Additional studies showed that optimal cross talk between FPR1 and CCR1 was dependent on the functional activity of protein kinase Cβ. These results provide a mechanistic basis for the capacity of certain GPCR ligands to exert rapid and selective cross-inactivation of other chemoattractant receptors, and suggest that FPR1 is able to exert "traffic control" in the migration of inflammatory cells by rapidly inhibiting the cell responses to potentially "low-priority" chemoattractants such as CCR1 agonists without inhibiting the response to "higher priority" CCR2 chemoattractants.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, ≥97% (HPLC)
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
O-Phospho-L-Serin
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, BioXtra, ≥99.0% (TLC)
Sigma-Aldrich
Fura 2-AM, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Fura 2-AM, ≥95% (HPLC)
Supelco
4-tert.-Octylphenol-monoethoxylat -Lösung, 10 μg/mL in acetone, analytical standard