Merck
SUMOylation occurs in acute kidney injury and plays a cytoprotective role.
Biochimica et biophysica acta (2014-12-24)
Chunyuan Guo, Qingqing Wei, Yunchao Su, Zheng Dong
ZUSAMMENFASSUNG
SUMOylation is a form of post-translational modification where small ubiquitin-like modifiers (SUMO) are covalently attached to target proteins to regulate their properties. SUMOylation has been demonstrated during cell stress and implicated in cellular stress response. However, it is largely unclear if SUMOylation contributes to the pathogenesis of kidney diseases, such as acute kidney injury (AKI). Here we have demonstrated a dynamic change of protein SUMOylation in ischemic and cisplatin nephrotoxic AKI in mice. In rat kidney proximal tubular cells (RPTC), cisplatin-induced SUMOylation was diminished by two antioxidants (N-acetylcysteine and dimethylurea), supporting a role of oxidative stress in the activation of SUMOylation. In addition, SUMOylation by SUMO-2/3, but not SUMO-1, was partially suppressed by pifithrin-alpha (a pharmacological inhibitor of p53), supporting a role of p53 in SUMOylation by SUMO-2/3. We further examined the role of SUMOylation during cisplatin treatment of RPTC by using ginkgolic acid (GA), a pharmacological inhibitor of SUMOylation. Pretreatment with GA suppressed SUMOylation and importantly, GA enhanced apoptosis during cisplatin incubation. Taken together, the results demonstrate the first evidence of SUMOylation in AKI and suggest that SUMOylation may play a cytoprotective role in kidney tubular cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Natriumazid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
N-Ethylmaleimid, purum p.a., ≥99.0% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumazid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
cis-Diamminplatin(II)-dichlorid, crystalline
Sigma-Aldrich
Natriumazid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
N-Ethylmaleimid, crystalline, ≥98% (HPLC)
Sigma-Aldrich
N-Ethylmaleimid, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Natriumazid, BioXtra
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diaminplatin(II)-dichlorid, ≥99.9% trace metals basis
Sigma-Aldrich
N-Ethylmaleimid, BioXtra, ≥98% (HPLC)
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Pifithrin-α, ≥95% (HPLC), powder
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
trans-Platin(II)diammindichlorid
Cisplatin Unreinheit A, European Pharmacopoeia (EP) Reference Standard
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard