Merck
  • Quantitative measurement of organic acids in tissues from gastric cancer patients indicates increased glucose metabolism in gastric cancer.
Quantitative measurement of organic acids in tissues from gastric cancer patients indicates increased glucose metabolism in gastric cancer.
PloS one (2014-06-10)
Hoon Hur, Man Jeong Paik, Yi Xuan, Duc-Toan Nguyen, In-Hye Ham, Jisoo Yun, Yong Kwan Cho, Gwang Lee, Sang-Uk Han
ZUSAMMENFASSUNG
The levels of organic acids representing metabolic pathway end products are important indicators of physiological status, and may be associated with metabolic changes in cancer. The aim of this study is to investigate the levels of organic acids in cancerous and normal tissues from gastric cancer patients and to confirm the role of metabolic alterations in gastric carcinogenesis. Organic acids in normal and cancerous tissues from forty-five patients with gastric adenocarcinoma were investigated by gas chromatography-mass spectrometry in selected ion monitoring mode as methoxime/tert-butyldimethylsilyl derivatives. We analysed the significant differences in the levels of organic acids in normal and cancer tissues and investigated the correlation of these levels in cancer tissues with clinicopathological features. The levels of Krebs cycle components, including α-ketoglutaric acid, succinic acid, fumaric acid, malic acid and oxaloacetic acid, were significantly increased in cancer tissues compared to normal tissues. In addition, the levels of glycolytic products, including pyruvic acid and lactic acid, as well as the levels of ketone bodies, including 3-hydroxybutyric acid, were also significantly increased in cancer tissues compared to normal tissues. The levels of ketone bodies in cancer tissues with differentiated histology and in intestinal-type cancer tissues were significantly increased. The organic acid profiling analysis described here may be a generally useful clinical tool for understanding the complexity of metabolic events in gastric adenocarcinoma, and organic acids may have potential as metabolic markers for the future discovery of diagnostic and therapeutic modalities.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Diethylether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Diethylether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Diethylether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Zitronensäure, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
tert-Butyl(chlor)dimethylsilan, reagent grade, 97%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Diethylether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
Brenztraubensäure, 98%
Sigma-Aldrich
Milchsäure, meets USP testing specifications
Sigma-Aldrich
Zitronensäure, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
O-Methylhydroxylamin -hydrochlorid, 98%
Sigma-Aldrich
Milchsäure, 88%, FCC
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
DL-Apfelsäure, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalessigsäure, ≥97% (HPLC)