Merck
Evidence of impairment of normal inflammatory reaction by a high-fat diet.
Genes and immunity (2014-02-28)
C Laflamme, G Bertheau-Mailhot, M S Giambelluca, N Cloutier, E Boilard, M Pouliot
ZUSAMMENFASSUNG
The murine dorsal air pouch model is a valuable tool for studying acute peripheral inflammatory reactions. We used this model to study the effect of diet on the onset of acute inflammation. Mice were fed a normal or a high-fat diet (HFD) for 5 weeks. Air pouches were raised and injected with non-stimulating (saline) or stimulating solution (saline containing lipopolysaccharides). After 4 h, leukocytes in the pouch fluid were enumerated, sorted and their viability measured. Cytokine/chemokine levels in the cell-free fluid were measured using a cytometric bead assay. Gene expression level was measured in leukocytes and in lining tissues using comparative real-time PCR. Leukocyte migration and cytokine/chemokine secretion were decreased substantially in mice fed the HFD. In contrast, leptin levels were elevated. Gene expression profiles in leukocytes recovered from the pouch and in the pouch-lining tissue (believed to have an important role in the initiation of granulocyte recruitment) were depressed. Genes encoding CC and CXC family chemokines were among the most negatively affected. These results suggest that a HFD can alter peripheral tissue activation as well as leukocyte recruitment and response, thereby affecting the development of an effective local immune response, which could have deleterious consequences.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.