Merck
Non-genomic actions of estradiol and 4-OH-tamoxifen on murine breast cancer cells.
Oncology reports (2014-10-24)
Diego Raffo, Osvaldo Pontiggia, Elisa Bal de Kier Joffé, Marina Simian
ZUSAMMENFASSUNG
Estrogens and tamoxifen do not only exert their effects at the genomic level, but also play a role at the cell membrane activating downstream signaling pathways. We recently characterized an estrogen receptor-positive epithelial murine breast cancer cell line, LM05-E. Utilizing this cell line and MCF-7 cells, we compared the non-genomic effects of estradiol and 4-OH-tamoxifen. We showed that, similar to estradiol, tamoxifen activated the MAPK/ERK 1/2 pathway; however, we did not find activation of PI3K/AKT by either estradiol or tamoxifen. Short-term treatments with estradiol stimulated, whereas tamoxifen inhibited cell proliferation. Using pharmacological inhibitors we showed that the effect of estradiol was mediated by the MAPK/ERK 1/2 pathway, but that inhibition of this pathway did not affect tamoxifen. Surprisingly, however, blocking of PI3K/AKT signaling interfered with the inhibitory effect of tamoxifen. Analysis of the involvement of the EGFR support previous findings that designate this receptor as a mediator of the non-genomic effects of estradiol; blocking EGFR also reverses the inhibitory effect of tamoxifen. Finally, matrix metalloproteinases (MMPs) were confirmed to be involved in the proliferative effect of estradiol. These results demonstrated the novel non-genomic effects of tamoxifen and revealed that pathways downstream of EGFR and PI3K/AKT are involved in the inhibition of cell proliferation. Caution should be exercised when analyzing strategies that aim at combining endocrine therapy with specific signaling inhibitors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
β-Estradiol, meets USP testing specifications
Supelco
Tamoxifen, analytical standard
Sigma-Aldrich
Ethanol
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
β-Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
PD 98,059, solid
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard