Merck
  • Optimization of ultrasonic-assisted extraction of continentalic acid from the root of Aralia continentalis by using the response surface methodology.
Optimization of ultrasonic-assisted extraction of continentalic acid from the root of Aralia continentalis by using the response surface methodology.
Archives of pharmacal research (2013-12-03)
Hyo Seon Kim, A Yeong Lee, Ji Eun Jo, Byeong Cheol Moon, Yunui Ji, Ho Kyoung Kim
ZUSAMMENFASSUNG
In order to optimize the extraction conditions of continentalic acid, the main compound from Aralia continentalis root, we developed a model using the response surface methodology. The continentalic acid yield was analyzed and quantified by high-performance liquid chromatography coupled with UV detection. The extraction solvent, temperature, and time the three main factors for ultrasound-assisted extraction were optimized using the central composite design. Analysis of variance showed a good model fit (R (2) = 0.9323). The maximum extraction of continentalic acid obtained experimentally was 1.0138 % under an extraction temperature of 33 °C and extraction time of 28 min when 100 % ethanol was used as the solvent. The experimental value was in good agreement (the yield 1.0103 %) with those predicted values. The results clearly showed that quality by statistical design could be effectively applied to optimize extraction of continentalic acid.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%