Merck
  • Development of an analytical method for the determination of the misuse in sports of boldenone through the analysis of urine by on-line coupling liquid chromatography-gas chromatography-combustion-isotope ratio mass spectrometry.
Development of an analytical method for the determination of the misuse in sports of boldenone through the analysis of urine by on-line coupling liquid chromatography-gas chromatography-combustion-isotope ratio mass spectrometry.
Journal of chromatography. A (2014-12-03)
R M Toledano, E M Díaz-Plaza, J M Cortes, A Aragón, A M Vázquez, J Villén, J Muñoz-Guerra
ZUSAMMENFASSUNG
Boldenone (Bo), androsta-1,4-dien-17β-ol-3-one, is an anabolic androgenic steroid not clinically approved for human application. Despite this, many cases are reported every year of athletes testing positive for Bo or its main metabolite 5β-androst-1-en-17β-ol-3-one (BoM). Recently the capability of different human intestinal bacteria to produce enzymes able to modify endogenous steroids in Bo has been demonstrated. When a urinary concentration of Bo and/or BoM between 5 and 30 ng/mL is measured a complementary analysis by gas chromatography combustion isotope ratio mass spectrometry (GC-C-IRMS) must be carried out to discriminate the endogenous or exogenous origin. In the present work, a novel analytical method that couples LC-GC by means of the TOTAD interface with C-IRMS is described. The method is based on a first RPLC separation of unacetyled steroids, followed by acetylation and automated on-line LC-GC-C-IRMS, which includes a second RPLC clean-up of acetyl Bo and BoM, isolation of the two fractions in a fraction collector and their consecutive analysis by GC-C-IRMS. The method has been applied to the analysis of urine samples fortified at 5 and 10 ng/mL, where it has shown a good performance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Pyridin, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Pyridin, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
Pentan, ≥99% (GC)
Sigma-Aldrich
Pentan, anhydrous, ≥99%
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Pyridin, ≥99%
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Pyridin, biotech. grade, ≥99.9%
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Supelco
Pyridin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Pyridin, analytical standard
Supelco
Pentan, analytical standard
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material