Merck
Direct drug analysis from oral fluid using medical swab touch spray mass spectrometry.
Analytica chimica acta (2015-02-24)
Valentina Pirro, Alan K Jarmusch, Marco Vincenti, R Graham Cooks
ZUSAMMENFASSUNG
Fourteen common drugs of abuse were identified in spiked oral fluid (ng mL(-1) levels), analyzed directly from medical swabs using touch spray mass spectrometry (TS-MS), exemplifying a rapid test for drug detection. Multiple stages of mass analysis (MS(2) and MS(3)) provided identification and detection limits sought by international forensic and toxicological societies, Δ(9)-THC and buprenorphine excluded. The measurements were made using a medical swab as both the sampling probe and means of ionization. The adaptation of medical swabs for TS-MS analysis allows non-invasive and direct sampling of neat oral fluid. Data acquisition was rapid, seconds per drug, and MS(3) ensured reliable identification of illicit drugs. The reported data were acquired to investigate (i) ionization of common drugs from commercial swabs, (ii) ion intensity over spray duration, and (iii) dynamic range, all as initial steps in development of a quantitative method. The approach outlined is intended for point-of-care drug testing using oral fluid in clinical applications as well as in situ settings, viz. in forensic applications. The proof-of-concept results presented will require extension to other controlled substances and refinement in analytical procedures to meet clinical/legal requirements.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Ameisensäure -Lösung, BioUltra, 1.0 M in H2O
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar