Merck
  • Combination therapy of prostate cancer with HPMA copolymer conjugates containing PI3K/mTOR inhibitor and docetaxel.
Combination therapy of prostate cancer with HPMA copolymer conjugates containing PI3K/mTOR inhibitor and docetaxel.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-12-08)
Yan Zhou, Jiyuan Yang, Rui Zhang, Jindřich Kopeček
ZUSAMMENFASSUNG
Combination therapies have been investigated to address the current challenges of anti-cancer therapeutics. In particular, a novel paradigm of combination therapy targeting both cancer stem/progenitor cells and bulk tumor cells is promising to improve the long-term therapeutic benefit against prostate cancer. Among the therapeutic agents with anti-CSC activities, the PI3K/mTOR inhibitors exhibit preferential inhibitory effect on prostate cancer stem/progenitor cells and potent cytotoxicity against bulk tumor cells. The combination of PI3K/mTOR inhibitor and traditional chemotherapy docetaxel may show superior therapeutic effect over single drug treatment. Aiming to further improve the combinational anti-tumor and anti-CSC effect, we developed the combination therapy containing two HPMA copolymer-drug conjugates, incorporated with PI3K/mTOR inhibitor GDC-0980 (P-(GDC-0980)) and docetaxel (P-DTX), respectively. The anti-tumor and anti-CSC effects of the single and combination therapy were investigated in vitro and on PC-3 prostate cancer xenografts in nude mice. Our evaluations showed that P-(GDC-0980) suppressed CD133+ prostate stem/progenitor cell growth even at the low dose which does not cause significant growth inhibition in bulk tumor cells. The combination therapy exhibited effective anti-CSC effect as well as enhanced anti-bulk tumor effect in vitro. Among all the single and combination dosing regimens of free drugs and conjugates, the macromolecular combination therapy showed significantly prolonged mice survival in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Supelco
Trifluoressigsäure, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Aphidicolin, ≥98% (HPLC), powder
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard