Merck
  • A comprehensive approach to in vitro functional evaluation of Ag/alginate nanocomposite hydrogels.
A comprehensive approach to in vitro functional evaluation of Ag/alginate nanocomposite hydrogels.
Carbohydrate polymers (2014-07-20)
Jasmina Stojkovska, Danijela Kostić, Željka Jovanović, Maja Vukašinović-Sekulić, Vesna Mišković-Stanković, Bojana Obradović
ZUSAMMENFASSUNG
In this work, we present a comprehensive approach to evaluation of alginate microbeads with included silver nanoparticles (AgNPs) at the concentration range of 0.3-5mM for potential biomedical use by combining cytotoxicity, antibacterial activity, and silver release studies. The microbeads were investigated regarding drying and rehydration showing retention of ∼ 80-85% of the initial nanoparticles as determined by UV-vis and SEM analyses. Both wet and dry microbeads were shown to release AgNPs and/or ions inducing similar growth delays of Staphylococcus aureus and Escherichia coli at the total released silver concentrations of ∼ 10 μg/ml. On the other hand, these concentrations were highly toxic for bovine chondrocytes in conventional monolayer cultures while nontoxic when cultured in alginate microbeads under biomimetic conditions in 3D perfusion bioreactors. The applied approach outlined directions for further optimization studies demonstrating Ag/alginate microbeads as potentially attractive components of soft tissue implants as well as antimicrobial wound dressings.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammoniumhydroxid -Lösung, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Salpetersäure, ACS reagent, 70%
Sigma-Aldrich
Silbernitrat, ACS reagent, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Ammoniumhydroxid -Lösung, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Kaliumnitrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Silbernitrat, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Salpetersäure, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Kaliumnitrat, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Kaliumnitrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Silbernitrat, 99.9999% trace metals basis
Sigma-Aldrich
Salpetersäure, red, fuming, HNO3 >90 %
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kaliumnitrat, BioReagent, suitable for cell culture, suitable for plant cell culture