Merck
  • The calcium-binding protein complex S100A8/A9 has a crucial role in controlling macrophage-mediated renal repair following ischemia/reperfusion.
The calcium-binding protein complex S100A8/A9 has a crucial role in controlling macrophage-mediated renal repair following ischemia/reperfusion.
Kidney international (2014-06-19)
Mark C Dessing, Alessandra Tammaro, Wilco P Pulskens, Gwendoline J Teske, Loes M Butter, Nike Claessen, Marco van Eijk, Tom van der Poll, Thomas Vogl, Johannes Roth, Sandrine Florquin, Jaklien C Leemans
ZUSAMMENFASSUNG
Upon ischemia/reperfusion (I/R)-induced injury, several damage-associated molecular patterns are expressed including the calcium-binding protein S100A8/A9 complex. S100A8/A9 can be recognized by Toll-like receptor-4 and its activation is known to deleteriously contribute to renal I/R-induced injury. To further test this, wild-type and S100A9 knockout mice (deficient for S100A8/A9 complex) were subjected to renal I/R. The expression of S100A8/A9 was significantly increased 1 day after I/R and was co-localized with Ly6G (mouse neutrophil marker)-positive cells. These knockout mice displayed similar renal dysfunction and damage and neutrophil influx compared with wild-type mice at this early time point. Interestingly, S100A9 knockout mice displayed altered tissue repair 5 and 10 days post I/R, as reflected by increased renal damage, sustained inflammation, induction of fibrosis, and increased expression of collagens. This coincided with enhanced expression of alternatively activated macrophage (M2) markers, while the expression of classically activated macrophage (M1) markers was comparable. Similarly, S100A9 deficiency affected M2, but not M1 macrophage polarization in vitro. During the repair phase following acute kidney injury, S100A9 deficiency affects M2 macrophages in mice leading to renal fibrosis and damage. Thus, S100A8/A9 plays a crucial part in controlling macrophage-mediated renal repair following I/R.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
SAFC
L-Glutamin
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Supelco
Aceton, analytical standard
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Lidocain, powder
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Lidocain, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis