Merck
Screening and comparison of antioxidant activities of polysaccharides from Coriolus versicolor.
International journal of biological macromolecules (2014-05-27)
Xiaowen Sun, Yanping Sun, Qingbo Zhang, Hongwei Zhang, Bingyou Yang, Zhibin Wang, Weiguo Zhu, Bin Li, Qiuhong Wang, Haixue Kuang
ZUSAMMENFASSUNG
Six polysaccharide fractions (Coriolus versicolor polysaccharides: CVPS-1, CVPS-2, CVPS-3, CVPS-4, CVPS-5 and CVPS-6) were isolated and purified from the fruiting bodies of C. versicolor by ion exchange chromatography and gel chromatography. Their chemical and physical characteristics were determined by chemical methods, high performance liquid chromatography, and high-performance gel-permeation chromatography. Finally, 1,1-diphenyl-2-picrylhydrazyl (DPPH) radical assay, superoxide radical assay, and hydroxyl radical assay were carried out to test the antioxidant activities of CVPS in vitro. The results indicated that the six CVPS fractions were acidic heteropolysaccharides, composed of mannose, rhamnose, glucuronic acid, glucose and fructose with different ratios. The molecular weights of CVPS-1, CVPS-2, CVPS-3, CVPS-4, CVPS-5 and CVPS-6 were 1740, 1480, 568, 880, 1260 and 1840kDa and the protein contents were 4.2%, 6.4%, 8.5%, 7.8%, 6.5% and 3.9%, respectively. Among the six fractions, CVPS with lower molecular weight, higher protein content and larger uronic acid amount, basically exhibited higher radical scavenging effects at the same concentration. Compared with other fractions, CVPS-3 exhibited the highest antioxidant activities. The effects of the molecular weight, protein content and uronic acid amount of the polysaccharides appeared to be significant on the improvement of the bioactivities.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Schwefelsäure, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Trichloressigsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trichloressigsäure -Lösung, 6.1 N
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
USP
Ascorbinsäure, United States Pharmacopeia (USP) Reference Standard
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Fruktose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol -Lösung, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Phenol, ≥99%
Supelco
L-Ascorbinsäure, analytical standard
Sigma-Aldrich
Trichloressigsäure, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Verflüssigtes Phenol, ≥89.0%