Merck
  • Simultaneous evaluation of human CYP3A4 and ABCB1 induction by reporter assay in LS174T cells, stably expressing their reporter genes.
Simultaneous evaluation of human CYP3A4 and ABCB1 induction by reporter assay in LS174T cells, stably expressing their reporter genes.
Biopharmaceutics & drug disposition (2014-11-21)
Keita Inami, Takamitsu Sasaki, Takeshi Kumagai, Kiyoshi Nagata
ZUSAMMENFASSUNG
The bioavailability of orally administered therapies are often significantly limited in the human intestine by the metabolic activities of cytochrome P450 3A4 (CYP3A4) and P-glycoprotein (P-gp). Predicting whether candidate compounds induce CYP3A4 and P-gp is a crucial stage in the drug development process, as drug-drug interactions may result in the induction of intestinal CYP3A4 and P-gp. However, the assay systems needed to evaluate both CYP3A4 and P-gp induction in the intestine are yet to be established. To address this urgent requirement, LS174T cells were used to create two stable cell lines expressing the CYP3A4 or ATP-binding cassette subfamily B member 1 (ABCB1, encoding P-gp) reporter genes. First, these stable cells were tested by treatment with 1α,25-dihydroxyvitamin D3 (1,25(OH)2D3), all-trans-retinoic acid (ATRA) and 9-cis-retinoic acid (9-cis RA) that induce CYP3A4 and P-gp in the intestines. All these compounds significantly increased both CYP3A4 and ABCB1 reporter activities in the stable cell lines. To simultaneously assess the induction of CYP3A4 and ABCB1, both stable cells were co-cultivated to measure their reporter activities. The mixed cells showed a significant increase in the CYP3A4 and ABCB1 reporter activities following treatment with 1,25(OH)2 D3, ATRA, and 9-cis RA. These activity levels were maintained after passaging more than 20 times and following multiple freeze-thaw cycles. These results demonstrate that our established cell lines can be used to evaluate simultaneously CYP3A4 and ABCB1 induction in the intestines, providing a valuable in vitro model for the evaluation of future drug candidates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Retinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dimethylsulfoxid, analytical standard
USP
Tretinoin, United States Pharmacopeia (USP) Reference Standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Tretinoin, European Pharmacopoeia (EP) Reference Standard
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®