Merck
  • Wip1-deficient neutrophils significantly promote intestinal ischemia/reperfusion injury in mice.
Wip1-deficient neutrophils significantly promote intestinal ischemia/reperfusion injury in mice.
Current molecular medicine (2015-01-21)
J Du, X Shen, Y Zhao, X Hu, B Sun, W Guan, S Li, Y Zhao
ZUSAMMENFASSUNG
Wip1 is a serine/threonine protein phosphatase which plays a critical role in neutrophil development and maturation. In the present study, we used a neutrophil-dependent model of intestinal ischemia/reperfusion (I/R) injury to identify the role of Wip1 in neutrophil function under the condition of oxidative stress and inflammation. Wip1- deficient mice displayed more severe intestinal I/R injury with increased infiltration of neutrophils and higher expression of chemokines like CXCL-1, CXCL-2 and CCL-2, as well as inflammatory cytokine like TNF-α and IL-17. Studies in Wip1KOa→WT full hematopoietic chimera mice showed that Wip1 intrinsically regulated the function of immune cells after intestinal I/R injury. Through adoptive transfer of neutrophils from WT mice or mice with deficiency of IL-17, IL-17/Wip1 or Wip1, we demonstrated that Wip1KO neutrophils produced more IL-17 and eventually led to more severe intestinal I/R injury. Thus, our findings identify Wip1 as an intrinsic negative regulator of neutrophil inflammation in intestinal I/R injury process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material