Merck
Cathepsin D repairing role in photodamaged skin barrier.
Skin pharmacology and physiology (2014-11-18)
Yue Zheng, Haiyan Chen, Wei Lai, Qingfang Xu, Chen Liu, Lin Wu, Howard I Maibach
ZUSAMMENFASSUNG
BACKGROUNDS/OBJECTIVES: Cathepsin D plays an important part in maintaining a normal skin barrier. Our previous study found that cathepsin D decreased in chronic photodamaged skin. This study investigated the cathepsin D content change in the stratum corneum (SC) and the repairing role of cathepsin D in chronic photodamaged skin barrier via the application of cathepsin D gel. Cathepsin D gel (0.001%) was applied to chronic photodamaged (sun-exposed forearm) human skin on identical sites (1 cm(2)/area) twice daily for 2 weeks. At 30 min and at 1, 3, 7, and 14 days, skin hydration and transepidermal water loss (TEWL) average values were detected via noninvasive skin detection equipment. Cathepsin D and transglutaminase (TGase)-1 in the skin sublayers were separated and detected via tape stripping, ELISA and Western blot. After 2 weeks of cathepsin D gel application, the skin moisture value increased from 86.8 ± 1.2 to 95.2 ± 2.7 (p < 0.05), while TEWL decreased from 17.88 ± 1.87 to 11.58 ± 2.14 (p < 0.05). Cathepsin D protein was detected in the upper epidermis (12.6 ± 2.6 ng/cm(2)), mid-epidermis (8.4 ± 0.8 ng/cm(2)) and deep epidermis (16.2 ± 2.6 ng/cm(2)) in the cathepsin D gel group compared to the control group (2.2 ± 0.7, 3.0 ± 1.1 and 3.85 ± 1.4 ng/cm(2), respectively; p < 0.05). TGase-1 enzyme expression was upregulated 2.54 ± 0.19 times in the matrix gel-treated skin. These data suggest that cathepsin D gel could increase the SC cathepsin D content and repair the epidermal barrier in chronic photodamaged skin. The mechanism might be related to increasing TGase-1 expression and activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zitronensäure, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Triethanolamin, ≥99.0% (GC)
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
Zitronensäure, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Triethanolamin, reagent grade, 98%
Supelco
Citronensäure, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Zitronensäure, BioUltra, anhydrous, ≥99.5% (T)
USP
Zitronensäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Zitronensäure, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Triethanolamin, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
2-Hydroxybuttersäure, 97%
Sigma-Aldrich
Triethanolamin, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Triethanolamin, puriss., meets analytical specification of NF, ≥99% (GC)
Supelco
Zitronensäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Zitronensäure, anhydrous, European Pharmacopoeia (EP) Reference Standard
Trolamin, European Pharmacopoeia (EP) Reference Standard
Supelco
Triethanolamin, analytical standard