Merck
  • Effect of A disintegrin and metalloproteinase 10 gene silencing on the proliferation, invasion and migration of the human tongue squamous cell carcinoma cell line TCA8113.
Effect of A disintegrin and metalloproteinase 10 gene silencing on the proliferation, invasion and migration of the human tongue squamous cell carcinoma cell line TCA8113.
Molecular medicine reports (2014-10-22)
Yuan Shao, Xiao-Ying Sha, Yan-Xia Bai, Fang Quan, Sheng-Li Wu
ZUSAMMENFASSUNG
The present study aimed to investigate the effect of A disintegrin and metalloproteinase 10 (ADAM10) gene silencing on the proliferation, migration and invasion of the human tongue squamous cell carcinoma cell line TCA8113. RNA interference was used to knock down the expression of ADAM10 in the TCA8113 cell line and the proliferation, migration and invasive ability of the treated cells were observed in vitro. The expression levels of epidermal growth factor receptor (EGFR) and E-cadherin in the treated cells were determined by western blot analysis. The proliferation, migration and invasion abilities of cells in the ADAM10 siRNA-treated group were significantly lower than those in the control groups (P<0.05). In addition, compared with the control groups, the expression levels of EGFR and E-cadherin in the ADAM10 siRNA-treated cells were significantly decreased (P<0.05) and increased (P<0.05), respectively. These results suggested that ADAM10 is important in regulating the proliferation, invasion and migration of the human tongue squamous cell carcinoma cell line TCA8113 and that the mechanism may, at least in part, be associated with the upregulation of EGFR and the downregulation of E-cadherin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®