Merck
  • Differentiation of degrees of ripeness of Catuai and Tipica green coffee by chromatographical and statistical techniques.
Differentiation of degrees of ripeness of Catuai and Tipica green coffee by chromatographical and statistical techniques.
Food chemistry (2014-12-23)
Samo Smrke, Ivana Kroslakova, Alexia N Gloess, Chahan Yeretzian
ZUSAMMENFASSUNG
The quality of green coffee is influenced by the degree of ripeness of the fruit at harvest. The aim of this study was to identify chemical markers differentiating between degrees of ripeness. Two coffee varieties, Catuai and Tipica, from the same farm were analysed using the following parameters and methods: caffeine and chlorogenic acid content using high-performance liquid chromatography (HPLC), sucrose content using hydrophilic interaction chromatography, high-molecular weight fraction (HMW) using high-performance size-exclusion chromatography (HPSEC) and volatile compounds using headspace solid phase micro extraction gas chromatography/mass spectrometry. The best method for differentiating between degrees of ripeness was found to be principal component analysis (PCA) based on HPLC data. HPSEC showed differences in the HMW fraction for different degrees of ripeness and both coffee varieties. Volatile profiles allowed separation of both varieties; yet, except for ripe Catuai, no separation was achieved for the degree of ripeness.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Supelco
Koffein, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Koffein, powder, ReagentPlus®
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumphosphat, 96%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture