Merck
  • Transcriptome analysis of hen preadipocytes treated with an adipogenic cocktail (DMIOA) with or without 20(S)-hydroxylcholesterol.
Transcriptome analysis of hen preadipocytes treated with an adipogenic cocktail (DMIOA) with or without 20(S)-hydroxylcholesterol.
BMC genomics (2015-03-15)
Alemu Regassa, Woo Kyun Kim
ZUSAMMENFASSUNG
20(S)-hydroxycholesterol (20(S)) potentially reduces adipogenesis in mammalian cells. The role of this oxysterol and molecular mechanisms underlying the adipogenesis of preadipocytes from laying hens have not been investigated. This study was conducted to 1. Analyze genes differentially expressed between preadipocytes treated with an adipogenic cocktail (DMIOA) containing 500 nM dexamethasone, 0.5 mM 3-isobutyl-1-methylxanthine, 20 μg/mL insulin and 300 μM oleic acid (OA) and control cells and 2. Analyze genes differentially expressed between preadipocytes treated with DMIOA and those treated with DMIOA + 20(S) using Affymetrix GeneChip® Chicken Genome Arrays. In experiment one, where we compared the gene expression profile of non-treated (control) cells with those treated with DMIOA, out of 1,221 differentially expressed genes, 755 were over-expressed in control cells, and 466 were over-expressed in cells treated with DMIOA. In experiment two, where we compared the gene expression profile of DMIOA treated cells with those treated with DMIOA+20(S), out of 212 differentially expressed genes, 90 were over-expressed in cells treated with DMIOA, and 122 were over-expressed in those treated with DMIOA+20(S). Genes over-expressed in control cells compared to those treated with DMIOA include those involved in cell-to-cell signaling and interaction (IL6, CNN2, ITGB3), cellular assembly and organization (BMP6, IGF1, ACTB), and cell cycle (CD4, 9, 38). Genes over-expressed in DMIOA compared to control cells include those involved in cellular development (ADAM22, ADAMTS9, FIGF), lipid metabolism (FABP3, 4 and 5), and molecular transport (MAP3K8, PDK4, AGTR1). Genes over-expressed in cells treated with DMIOA compared with those treated with DMIOA+20(S) include those involved in lipid metabolism (ENPP2, DHCR7, DHCR24), molecular transport (FADS2, SLC6A2, CD36), and vitamin and mineral metabolism (BCMO1, AACS, AR). Genes over-expressed in cells treated with DMIOA+20(S) compared with those treated with DMIOA include those involved in cellular growth and proliferation (CD44, CDK6, IL1B), cellular development (ADORA2B, ATP6VOD2, TNFAIP3), and cell-to-cell signaling and interaction (VCAM1, SPON2, VLDLR). We identified important adipogenic regulators and key pathways that would help to understand the molecular mechanism of the in vitro adipogenesis in laying hens and demonstrated that 20(S) is capable of suppressing DMIOA-induced adipogenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Adenosin, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Adenosin, BioReagent, suitable for cell culture
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
tert-Butylacetoacetat, reagent grade, 95%
Sigma-Aldrich
Adenosin
Supelco
Adenosin, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.
Adenosin, European Pharmacopoeia (EP) Reference Standard