Merck
  • Quantitative LC-MS/MS determination of flupirtine, its N-acetylated and two mercapturic acid derivatives in man.
Quantitative LC-MS/MS determination of flupirtine, its N-acetylated and two mercapturic acid derivatives in man.
Journal of pharmaceutical and biomedical analysis (2014-12-03)
Eberhard Scheuch, Karen Methling, Patrick J Bednarski, Stefan Oswald, Werner Siegmund
ZUSAMMENFASSUNG
The non-opiate analgesic drug flupirtine was shown in vitro to undergo hydrolysis followed by N-acetylation to form D13223, glucuronidation and conjugation with glutathione to form the stable mercapturic acid derivatives M-424 and M-466. To quantify flupirtine and its metabolites in samples obtained in a clinical study in healthy subjects selected on their genotype of NAT2, UGT1A1 and GSTP1, two LC-MS/MS methods were developed. The validation range for flupirtine and D-13223 in serum was 0.5-500 ng/ml. For urine and feces, the validation ranges for flupirtine and D-13223 were 20-5000 ng/ml and 5.0-5000 ng/ml, respectively. M-424 and M-466 could be quantified in urine between 5.0 and 5000 ng/ml. Free flupirtine and D-13223 were separated from serum, urine and feces with liquid-liquid extraction. For flupirtine and D-13223, the chromatography was performed on a XTerra C18 column isocratically with a mobile phase consisting of ammonium formate buffer (pH 3.5mM) and acetonitrile (50:50; v/v), for M-466 and M-424 a Synergi(®) Fusion-RP column was used and a linear gradient method with water/HCOOH (pH 3) and acetonitrile. The mass spectrometer operated both with electro spray ionization in positive multiple reaction monitoring mode. The developed methods fulfilled the current FDA criteria on bioanalytical method validation for accuracy (error: -16.9 to 11.2%), precision (1.2-13.4%), recovery, stability and matrix effects over the observed analytical range. Thus, the methods were suitable to quantify flupirtine absorption and metabolic disposition in man after single intravenous and oral dosing (100mg) and repeated oral administration (400mg once daily).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Supelco
Ammoniumformat, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammoniumformat, reagent grade, 97%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ammoniumformat, ≥99.995% trace metals basis
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammoniumformiat -Lösung, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammoniumformat, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG