Merck
  • Retinoic acid signalling regulates the development of tonotopically patterned hair cells in the chicken cochlea.
Retinoic acid signalling regulates the development of tonotopically patterned hair cells in the chicken cochlea.
Nature communications (2014-05-23)
Benjamin R Thiede, Zoë F Mann, Weise Chang, Yuan-Chieh Ku, Yena K Son, Michael Lovett, Matthew W Kelley, Jeffrey T Corwin
ZUSAMMENFASSUNG
Precise frequency discrimination is a hallmark of auditory function in birds and mammals and is required for distinguishing similar sounding words, like 'bat,' 'cat' and 'hat.' In the cochlea, tuning and spectral separation result from longitudinal differences in basilar membrane stiffness and numerous individual gradations in sensory hair cell phenotypes, but it is unknown what patterns the phenotypes. Here we used RNA-seq to compare transcriptomes from proximal, middle and distal regions of the embryonic chicken cochlea, and found opposing longitudinal gradients of expression for retinoic acid (RA)-synthesizing and degrading enzymes. In vitro experiments showed that RA is necessary and sufficient to induce the development of distal-like hair cell phenotypes and promotes expression of the actin-crosslinking proteins, Espin and Fscn2. These and other findings highlight a role for RA signalling in patterning the development of a longitudinal gradient of frequency-tuned hair cell phenotypes in the cochlea.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Citral, 95%
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Citral, natural, ≥96%, FCC, FG
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Citral, mixture of cis and trans, ≥96%, FG
Supelco
Citral, analytical standard
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
BMS 493, ≥98% (HPLC)
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard