Merck
  • Rapid fluorescent detection of Escherichia coli K88 based on DNA aptamer library as direct and specific reporter combined with immuno-magnetic separation.
Rapid fluorescent detection of Escherichia coli K88 based on DNA aptamer library as direct and specific reporter combined with immuno-magnetic separation.
Journal of fluorescence (2014-04-26)
Zhihui Peng, Min Ling, Yi Ning, Le Deng
ZUSAMMENFASSUNG
Nucleic acid aptamers have long demonstrated the capacity to bind cells with high affinity so that they have been utilized to diagnose various important pathogens. In this study, a DNA aptamer library was on initial efforts developed to act as a specific reporter for rapid detection of enter toxigenic Escherichia coli (ETEC) K88 combined with immuno-magnetic separation (IMS). During a Whole-cell Systematic Evolution of Ligands by Exponential Enrichment (CELL-SELEX) procedure, the last selection pool against ETEC K88, which is named "DNA aptamer library" here, was selected and subsequently identified by flow cytometric analysis and confocal imaging. A K88 monoclonal antibody (mAb) with high affinity (K(aff): 1.616 ± 0.033 × 10(8) M(-1)) against K88 fimbrial protein was prepared, biotinylated and conjugated to streptavidin-coated magnetic beads (MBs). After the bacteria were effectively captured and enriched from the complex sample by immuno-magnetic beads (IMBs), 5'-FITC modified aptamer library was directly bound to target cells as a specific reporter for its detection. The detection system showed clearly high specificity and sensitivity with the detection limit of 1.1 × 10(3) CFU/ml in pure culture and 2.2 × 10(3) CFU/g in artificially contaminated fecal sample. The results also indicated that fluorophore-lablled DNA aptamer library as specific reporter could generate more reliable signals than individual aptamer with best affinity against target cells and implied it would have great applied potential in directly reporting bacteria from complex samples combined with IMS technology.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
(+)-Biotin-N-Hydroxysuccinimid-Ester, ≥98% (HPLC), powder
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis