Merck
  • Highly class-selective solid-phase extraction of bisphenols in milk, sediment and human urine samples using well-designed dummy molecularly imprinted polymers.
Highly class-selective solid-phase extraction of bisphenols in milk, sediment and human urine samples using well-designed dummy molecularly imprinted polymers.
Journal of chromatography. A (2014-08-19)
Xiaoli Sun, Jincheng Wang, Yun Li, Jing Jin, Jiajia Yang, Fang Li, Syed Mazhar Shah, Jiping Chen
ZUSAMMENFASSUNG
Dummy molecularly imprinted polymers (DMIPs) towards bisphenols (BPs) were prepared employing 1,1,1-tris(4-hydroxyphenyl)ethane (THPE) and phenolphthalein (PP) as dummy templates. The selectivity of the resulting DMIPs was evaluated by high-performance liquid chromatography (HPLC). Both PP-DMIP and THPE-DMIP showed excellent class selectivity towards bisphenols. THPE-DMIP prepared using the template molecule with three hydroxyphenyl functionalities achieved higher imprinting factors (IF) for the bisphenols over a range of 7.9-19.8. An efficient approach based on dummy molecularly imprinted solid phase extraction (DMISPE) coupled with HPLC-DAD was developed for selective extraction of eight bisphenols in sediment, milk and human urine samples using THPE-DMIP as sorbents. The method showed good recoveries (82-102%) and precision (RSD 0.2-4%, n=3) for these samples spiked at two concentration levels (25 and 250ngg(-1) or ngmL(-1)). The detection limits ranged between 0.6 and 1.1ngg(-1) or ngmL(-1). Efficient removal of sample matrix and interferences was also achieved for these samples after DMISPE process. The results demonstrated great potential of the optimized methods for sample preparation in the routine analysis of trace BPs in complex samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
4-Vinylpyridin, contains 100 ppm hydroquinone as inhibitor, 95%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
2,4,6-Trichlorphenol, 98%
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard