Merck
  • Exposure to a continuous low dose of tetrachlorodibenzo-p-dioxin impairs the development of the tooth root in lactational rats and alters the function of apical papilla-derived stem cells.
Exposure to a continuous low dose of tetrachlorodibenzo-p-dioxin impairs the development of the tooth root in lactational rats and alters the function of apical papilla-derived stem cells.
Archives of oral biology (2014-12-03)
Hao Guo, Liqiang Zhang, Kewen Wei, Jiangdong Zhao, Yurui Wang, Fang Jin, Kun Xuan
ZUSAMMENFASSUNG
Ubiquitous environmental pollutants such as 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) cause abnormalities in reproduction and development. TCDD inhibits the development of teeth, and its effects depend on its dose and the developmental stage of the tooth. Our aim here was to investigate the effect of lower doses of TCDD on the development of the tooth root in vivo and in vitro. We observed tooth root development in lactational rats exposed to continuous low doses of TCDD starting on postnatal day 6 using Mico-CT analyses and histopathological examinations. And then the characteristics of stem cells derived from the apical papilla (SCAPs) were evaluated and compared with SCAPs induced by lower doses of TCDD both in vitro and in vivo. The results of experiments showed that rat pups exposed to low dose TCDD at prenatal stage developed, dentine hypoplasia, and hypomineralization. Further, TCDD impaired the functions of SCAPs in vivo by inhibiting cell proliferation and osteogenic and odontogenic differentiation. The impairment of SCAPs after TCDD exposure was accompanied by increased expression of AHR, down-regulation of the expression of Runx2, and alkaline phosphatase, suggesting that the AHR pathway mediated the effects of TCDD. These results provide the first insights into the toxicity of TCDD, which adversely affects the development of the tooth root through indirectly altering the function of SCAPs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
USP
Ascorbinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
SAFC
L-Glutamin
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Supelco
L-Ascorbinsäure, analytical standard
Sigma-Aldrich
Propidiumjodid -Lösung