Merck
  • PCL/PEG core/sheath fibers with controlled drug release rate fabricated on the basis of a novel combined technique.
PCL/PEG core/sheath fibers with controlled drug release rate fabricated on the basis of a novel combined technique.
International journal of pharmaceutics (2014-04-23)
Hui Yu, Yongtang Jia, Chaoming Yao, Yanxia Lu
ZUSAMMENFASSUNG
A novel core/sheath fiber preparation method, which included the processes of blend electrospinning to produce the core fiber and UV-induced graft polymerization to fabricate the outer polymeric shell, was presented to provide designated fibers with different shell thicknesses. A hydrophilic drug, salicylic acid (SA), was loaded in the representative poly(ϵ-caprolactone) (PCL)/polyethylene glycol (PEG) core/sheath fibers, performed according to this combined technique. FTIR analysis indicated that the existence of hydrogen bonds between SA and the PCL matrix improved drug compatibility. Field emission scanning electron microscopy (FESEM) images indicated that the morphology and the diameter distribution of fibers changed significantly after the graft polymerization procedure. All the core/sheath fibers became more flexible and thicker compared with the core fiber. The water contact angle (WCA) test also noted the differences of these two fibers: PCL/PEG core/sheath fibers with cross-linked PEG surface exhibited more hydrophilic property. Moreover, in vitro SA release tests were conducted to explore the relationship between the PEG shell thickness and the drug release rate. A typical biphasic release mechanism was observed for the PCL/PEG core/sheath fibers, and their sustained release rates were controlled by the PEG shell thickness in a linear correlation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
Salicylsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzophenon, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Salicylsäure, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
N,N-Dimethylformamid, biotech. grade, ≥99.9%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dichlormethan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
USP
Salicylsäure, United States Pharmacopeia (USP) Reference Standard