Merck
  • Simple and sensitive screening and quantitative determination of 88 psychoactive drugs and their metabolites in blood through LC-MS/MS: application on postmortem samples.
Simple and sensitive screening and quantitative determination of 88 psychoactive drugs and their metabolites in blood through LC-MS/MS: application on postmortem samples.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-09-15)
Cristina Sempio, Luca Morini, Claudia Vignali, Angelo Groppi
ZUSAMMENFASSUNG
The aim of the study was to develop and validate a simple, sensitive and specific method for the detection and quantitative determination of 88 substances among psychoactive drugs and their metabolites in whole blood, and to apply the procedure to postmortem cases. Samples were consecutively diluted with methanol, acetonitrile and mobile phase. All the molecules were separated and then identified through a liquid chromatographic, tandem mass spectrometric system, and eventually fully validated according to the international guidelines. The method proved to be highly sensitive and specific and all the validation parameters fulfilled the acceptance criteria. In particular linearity was studied in the range LOQ-1000 ng/mL; matrix effects and carry over were negligible and the majority of the compounds assessed to be stable over several freeze and thaw processes. Olanzapine is the most unstable compound. Protryptiline and flupenthixol did not fulfilled acceptance criteria, and although their transitions were kept on the instrumental settings, they were not considered for the fully validation. The method was applied to several postmortem cases, and the results were compared to the GC-MS systematic toxicological analysis currently in use in our laboratory, assessing to be a good complementary procedure and providing a better sensitivity. The LC-MS/MS method could be easily applicable to routine analyses of postmortem samples, as well as to a screening procedure for clinical purposes; however it should be carried out in combination with a general unknown screening method.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Clozapin
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Haloperidol, powder
Supelco
Acetonitril, analytical standard