Merck
  • ER stress induced by the OCH1 mutation triggers changes in lipid homeostasis in Kluyveromyces lactis.
ER stress induced by the OCH1 mutation triggers changes in lipid homeostasis in Kluyveromyces lactis.
Research in microbiology (2015-01-13)
Elena Zanni, Giuseppe Maulucci, Donatella Pomata, Francesca Buiarelli, Ewa K Krasnowska, Tiziana Parasassi, Marco De Spirito, Hermann J Heipieper, Daniela Uccelletti
ZUSAMMENFASSUNG
In Kluyveromyces lactis yeast, OCH1 encodes for the α-1,6-mannosyltrasferase that adds the initial α-1,6-mannose to the outer-chains of N-glycoproteins. Kloch1-1 mutant cells showed altered calcium homeostasis and endoplasmic reticulum (ER) stress. Since ER plays a major role in lipid biosynthesis and lipid droplet (LD) formation, herein the impact of Och1p depletion on lipid homeostasis was investigated. Transcriptional profiles of genes involved in biosynthesis of fatty acids, their amount and composition changed in mutant cells. An increased amount of ergosterol was determined in these cells. Enhanced transcription of genes involved in both synthesis and mobilization of LDs was also found in Kloch1-1 cells, accompanied by a reduced amount of LDs. We provide evidence that ER alterations, determined by protein misfolding as a result of reduced N-glycosylation, induced altered lipid homeostasis in Kloch1-1 cells. Chemical chaperone 4-phenyl butyrate (4-PBA) slightly alleviated the LD phenotype in cells depleted of Och1p. Remarkably, complete suppression of ER stress, via increased expression of plasma membrane calcium channel subunit Mid1, fully restored lipid homeostasis in mutant cells. To further reinforce this finding, low numbers of LDs were observed in wild type cells when ER stress was triggered by DTT treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Fluorescein, free acid