Merck
  • Simultaneous determination of liensinine, isoliensinine and neferine in rat plasma by UPLC-MS/MS and application of the technique to pharmacokinetic studies.
Simultaneous determination of liensinine, isoliensinine and neferine in rat plasma by UPLC-MS/MS and application of the technique to pharmacokinetic studies.
Journal of ethnopharmacology (2015-02-01)
Guoxin Hu, Ren-ai Xu, Yao-yao Dong, Yi-yan Wang, Wen-wen Yao, Zhi-chuan Chen, Dongxin Chen, Tiao Bu, Ren-shan Ge
ZUSAMMENFASSUNG
The in vivo effects of traditional herbal medicines are generally mediated by multiple bioactive components. The main constituents of Lotus Plumule are alkaloids such as liensinine, isoliensinine and neferine. In this study, a simple, sensitive, and robust analytical method based on ultra-performance liquid chromatography tandem mass spectrometry (UPLC-MS/MS) has been developed for the determination of the three alkaloids in rat plasma using carbamazepine as internal standard (IS). After precipitation of the proteins with acetonitrile, chromatography was performed on an Acquity UPLC BEH C18 column (2.1mm×50mm, 1.7μm particle size) using a gradient elution with 0.1% formic acid in water and acetonitrile. Mass spectrometry involved positive electrospray ionization and multiple reaction monitoring (MRM) of the transitions at m/z 611.7→206.2 for liensinine, 611.3→192.2 for isoliensinine, 625.2→206.1 for neferine and m/z 237.1→194.2 for IS. The method was linear over the concentration range 5-1000ng/mL with a lower limit of quantifof 5ng/mL for each alkaloid. Inter- and intra-day precision (RSD%) were all within 11.4% and the accuracy (RE%) were equal or lower than 10.4%. Recoveries were more than 75.3% and matrix effects were not significant. Stability studies showed that the three alkaloids were stable under a variety of storage conditions. The method was successfully applied to a pharmacokinetic study involving intravenous administration of liensinine, isoliensinine and neferine to rats.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Ameisensäure -Lösung, BioUltra, 1.0 M in H2O