Merck
  • Determination of caprolactam and residual vinyl caprolactam monomer in soluplus by mixed mode gel permeation chromatography.
Determination of caprolactam and residual vinyl caprolactam monomer in soluplus by mixed mode gel permeation chromatography.
Journal of chromatographic science (2013-05-17)
Ashish Soman, Madhuri Jerfy
ZUSAMMENFASSUNG
Soluplus, a graft copolymer of polyethylene glycol, vinyl caprolactam and vinyl acetate, is designed to solubilize poorly soluble active pharmaceutical ingredients. A straightforward aqueous gel permeation chromatography method that exploits both size exclusion and adsorption modes of separation was used to separate and quantify the related residual vinyl caprolactam monomer and caprolactam impurity present in Soluplus. This methodology offers a single step analysis of caprolactam and the residual vinyl caprolacatam monomer, yielding similar results to reversed-phase chromatography measurements, which are time-consuming and may involve multi-step sample preparation. The results of this study demonstrate that gel permeation chromatography provides a viable option to traditional reversed-phase chromatography in the quantitative analysis of residual caprolactam and vinyl caprolactam monomers and can be extended to other monomer-polymeric systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
ε-Caprolactam, 99%
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Supelco
ε-Caprolactam, analytical standard
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material