Merck
  • Identification of glutathione conjugates of acetylene-containing positive allosteric modulators of metabotropic glutamate receptor subtype 5.
Identification of glutathione conjugates of acetylene-containing positive allosteric modulators of metabotropic glutamate receptor subtype 5.
Drug metabolism and disposition: the biological fate of chemicals (2015-01-31)
Xiaoliang Zhuo, Xiaohua Stella Huang, Andrew P Degnan, Lawrence B Snyder, Fukang Yang, Hong Huang, Yue-Zhong Shu, Benjamin M Johnson
ZUSAMMENFASSUNG
A recent medicinal chemistry campaign to identify positive allosteric modulators (PAMs) of metabotropic glutamate receptor subtype 5 (mGluR5) led to the discovery of potent compounds featuring an oxazolidinone structural core flanked by biaryl acetylene and haloaryl moieties. However, biotransformation studies of some of these mGluR5 PAMs demonstrated the formation of glutathione (GSH) conjugates. The conjugates in question were formed independently of NADPH as the main products in liver microsomes and liver cytosol (rat and human) and exhibited masses that were 307 u greater than their respective substrates, indicating the involvement of a reductive step in the formation of these metabolites. To further characterize the relevant metabolic sequences, GSH conjugates of (4R,5R)-5-(3-fluorophenyl)-4-(5-(pyrazin-2-ylethynyl)pyridin-3-yl)oxazolidin-2-one and (4R,5R)-5-(4-fluorophenyl)-4-(6-((3-fluoropyridin-2-yl)ethynyl)pyridin-2-yl)oxazolidin-2-one were biosynthesized and isolated. Subsequent analysis by NMR showed that GSH had reacted with the acetylene carbon atoms of these mGluR5 PAMs, suggesting a conjugate addition mechanism and implicating cytosolic and microsomal GSH S-transferases (GSTs) in catalysis. Interestingly, five closely related mGluR5 PAMs were not similarly prone to the formation of GSH conjugates in vitro. These compounds also featured acetylenes, but were flanked by either phenyl or cyclohexyl rings, which indicated that the formation of GSH conjugates was influenced by proximal functional groups that modulated the electron density of the triple bond and/or differences in enzyme-substrate specificity. These results informed an ongoing drug-discovery effort to identify mGluR5 PAMs with drug-like properties and a low risk of reactivity with endogenous thiols.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Kaliumphosphat, tribasisch, reagent grade, ≥97%
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Supelco
Glutathion, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Glutathion, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ameisensäure -Lösung, BioUltra, 1.0 M in H2O
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar