Merck
  • Umbilical cord mesenchymal stem cells inhibit the differentiation of circulating T follicular helper cells in patients with primary Sjögren's syndrome through the secretion of indoleamine 2,3-dioxygenase.
Umbilical cord mesenchymal stem cells inhibit the differentiation of circulating T follicular helper cells in patients with primary Sjögren's syndrome through the secretion of indoleamine 2,3-dioxygenase.
Rheumatology (Oxford, England) (2014-08-30)
Rui Liu, Dinglei Su, Min Zhou, Xuebing Feng, Xia Li, Lingyun Sun
ZUSAMMENFASSUNG
The aim of this study was to investigate the effect of umbilical cord mesenchymal stem cells (UC-MSCs) on circulating T follicular helper (cTfh) cells in primary SS (pSS) patients. The percentage of CXCR5(+)PD-1(+)CD4(+) T cells in peripheral blood mononuclear cells (PBMCs) was analysed by flow cytometry. PBMCs were co-cultured with UC-MSCs by cell-to-cell contact or in a trans-well system. Naive CD4(+) T cells were isolated from PBMCs and then co-cultured with UC-MSCs under Tfh cell-polarizing conditions. The percentage of CXCR5(+)PD-1(+)CD4(+) T cells, carboxyfluorescein succinimidyl ester (CFSE) fluorescence intensity and annexin V were determined by flow cytometric analysis. Real-time PCR and Luminex cytokine assay were performed to detect mRNA expression and supernatant protein levels. The activity of indoleamine 2,3-dioxygenase (IDO) was measured by HPLC. Increased frequency of cTfh cells was found in pSS patients and was positively correlated with serum anti-La/SSB levels and the European League Against Rheumatism SS Disease Activity Index score. In vitro, UC-MSCs suppressed the differentiation and proliferation of cTfh cells. Real-time PCR analysis showed significantly higher IDO mRNA expression on UC-MSCs when co-cultured with naive CD4(+) T cells under Tfh cell-polarizing conditions in pSS patients. However, IDO mRNA expression on UC-MSCs was only a little higher when UC-MSCs were co-cultured with naive CD4(+) T cells in a trans-well system. In addition, HPLC showed increased IDO enzymic activity in the supernatant of UC-MSCs co-cultured with naive CD4(+) T cells under Tfh cell-polarizing conditions in pSS. The addition of the IDO inhibitor 1-MT partly reversed the suppressive effect of UC-MSCs on the differentiation of cTfh cells. These results suggest an inhibitory effect of UC-MSCs on the differentiation of cTfh cells via the secretion of IDO, and soluble factors secreted by activated CD4(+) T cells might contribute to IDO secretion by UC-MSCs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
5(6)-Carboxyfluoreszeindiacetat-N-succinimidylester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Supelco
Calcium Standard für AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%