Merck
Heparin octasaccharide decoy liposomes inhibit replication of multiple viruses.
Antiviral research (2015-02-01)
Gabriel L Hendricks, Lourdes Velazquez, Serena Pham, Natasha Qaisar, James C Delaney, Karthik Viswanathan, Leila Albers, James C Comolli, Zachary Shriver, David M Knipe, Evelyn A Kurt-Jones, Deborah K Fygenson, Jose M Trevejo, Jennifer P Wang, Robert W Finberg
ZUSAMMENFASSUNG
Heparan sulfate (HS) is a ubiquitous glycosaminoglycan that serves as a cellular attachment site for a number of significant human pathogens, including respiratory syncytial virus (RSV), human parainfluenza virus 3 (hPIV3), and herpes simplex virus (HSV). Decoy receptors can target pathogens by binding to the receptor pocket on viral attachment proteins, acting as 'molecular sinks' and preventing the pathogen from binding to susceptible host cells. Decoy receptors functionalized with HS could bind to pathogens and prevent infection, so we generated decoy liposomes displaying HS-octasaccharide (HS-octa). These decoy liposomes significantly inhibited RSV, hPIV3, and HSV infectivity in vitro to a greater degree than the original HS-octa building block. The degree of inhibition correlated with the density of HS-octa displayed on the liposome surface. Decoy liposomes with HS-octa inhibited infection of viruses to a greater extent than either full-length heparin or HS-octa alone. Decoy liposomes were effective when added prior to infection or following the initial infection of cells in vitro. By targeting the well-conserved receptor-binding sites of HS-binding viruses, decoy liposomes functionalized with HS-octa are a promising therapeutic antiviral agent and illustrate the utility of the liposome delivery platform.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Piperidin, ReagentPlus®, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Piperidin, ≥99.5%, purified by redistillation
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Piperidin, biotech. grade, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamin, ≥97.0% (TLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamin, ≥99.0% (10 mg phospholipid per ml CHCl3, TLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Piperidin -Lösung, suitable for peptide synthesis, 20% in DMF
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-α-Phosphatidylethanolamin, Dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-RSV Antibody, glycoprotein, all type A, B strains, clone 131-2G, clone 131-2G, from mouse
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Piperidin, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.