Merck
  • Simultaneous quantitative determination of microcystin-LR and its glutathione metabolites in rat liver by liquid chromatography-tandem mass spectrometry.
Simultaneous quantitative determination of microcystin-LR and its glutathione metabolites in rat liver by liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-06-16)
Xiaochun Guo, Ping Xie, Jun Chen, Xun Tuo, Xuwei Deng, Shangchun Li, Dezhao Yu, Cheng Zeng
ZUSAMMENFASSUNG
The roles of glutathione (GSH) and cysteine (Cys) in the detoxification of Microcystin-LR (MC-LR) have recently become a popular area of research. However, lacking analysis methods for MC-LR-GSH and MC-LR-Cys (two main GSH pathway metabolites) in mammals, elucidation of the detoxification mechanism and metabolic pathway of MC-LR in mammals is difficult. In this study, a novel method for the simultaneous quantitative analysis of MC-LR, MC-LR-GSH and MC-LR-Cys in rat liver was developed and validated. The analytes were simultaneously extracted from rat liver using 3M sodium chloride solution containing 0.01M EDTA-Na2-5% acetic acid, followed by solid-phase extraction (SPE) on Oasis HLB and silica cartridges and determination by liquid chromatography-electrospray ionization mass spectrometry (LC-ESI-MS/MS). Under the optimized pretreatment conditions and instrument parameters, good recoveries of MC-LR, MC-LR-GSH and MC-LR-Cys were obtained at three concentrations (0.2, 1.0 and 2.5 μg g(-1) dry weight (DW)) with values ranging from 97.7 ± 4.2 to 98.7 ± 5.1%, 70.1 ± 4.8 to 71.1 ± 4.1% and 79.8 ± 3.5 to 81.4 ± 4.0%, respectively. The relative standard deviations (RSDs) of these compounds at 0.2, 1.0 and 2.5 μg g(-1) DW were between 4.3% and 6.9%. The limits of detection (LODs) were 0.005, 0.007 and 0.006 μg g(-1) DW and the limits of quantification (LOQs) were 0.017, 0.023 and 0.020 μg g(-1) DW for MC-LR, MC-LR-GSH and MC-LR-Cys, respectively. Furthermore, this method was successfully applied to both time- and dosage-effect studies of MC-LR, MC-LR-GSH and MC-LR-Cys in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
L-Cystein, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
L-Cystein, 97%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
L-Cystein, BioUltra, ≥98.5% (RT)
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard