Merck
MicroRNA-188 suppresses G1/S transition by targeting multiple cyclin/CDK complexes.
Cell communication and signaling : CCS (2014-10-12)
Jiangbin Wu, Qing Lv, Jie He, Haoxiang Zhang, Xueshuang Mei, Kai Cui, Nunu Huang, Weidong Xie, Naihan Xu, Yaou Zhang
ZUSAMMENFASSUNG
Accelerated cell cycle progression is the common feature of most cancers. MiRNAs can act as oncogenes or tumor suppressors by directly modulating cell cycle machinery. It has been shown that miR-188 is upregulated in UVB-irradiated mouse skin and human nasopharyngeal carcinoma CNE cells under hypoxic stress. However, little is known about the function of miR-188 in cell proliferation and growth control. Overexpression of miR-188 inhibits cell proliferation, tumor colony formation and G1/S cell cycle transition in human nasopharyngeal carcinoma CNE cells. Using bioinformatics approach, we identify a series of genes regulating G1/S transition as putative miR-188 targets. MiR-188 inhibits both mRNA and protein expression of CCND1, CCND3, CCNE1, CCNA2, CDK4 and CDK2, suppresses Rb phosphorylation and downregulates E2F transcriptional activity. The expression level of miR-188 also inversely correlates with the expression of miR-188 targets in human nasopharyngeal carcinoma (NPC) tissues. Moreover, studies in xenograft mouse model reveal that miR-188 is capable of inhibiting tumor initiation and progression by suppressing target genes expression and Rb phosphorylation. This study demonstrates that miR-188 exerts anticancer effects, via downregulation of multiple G1/S related cyclin/CDKs and Rb/E2F signaling pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete, EDTA-freier Proteasehemmer-Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Roche
Zellproliferation ELISA, BrdU (kolorimetrisch), sufficient for ≤1,000 tests
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Hydroxyharnstoff, 98%, powder
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Supelco
Harnstoff, analytical standard
Sigma-Aldrich
Harnstoff, meets USP testing specifications
USP
Harnstoff, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O
Millipore
Harnstoff -Lösung, suitable for microbiology, 40% in H2O
Harnstoff, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Harnstoff-12C, 99.9 atom % 12C
Hydroxycarbamid, European Pharmacopoeia (EP) Reference Standard