Merck
  • "Center punch" and "whole spot" bioanalysis of apixaban in human dried blood spot samples by UHPLC-MS/MS.
"Center punch" and "whole spot" bioanalysis of apixaban in human dried blood spot samples by UHPLC-MS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-03-10)
Naiyu Zheng, Long Yuan, Qin C Ji, Heidi Mangus, Yan Song, Charles Frost, Jianing Zeng, Anne-Françoise Aubry, Mark E Arnold
ZUSAMMENFASSUNG
Apixaban (Eliquis™) was developed by Bristol-Myers Squibb (BMS) and Pfizer to use as an antithrombotic/anticoagulant agent and has been recently approved for the prevention of stroke and systemic embolism in patients with nonvalvular atrial fibrillation. A clinical study of apixaban, sponsored by BMS and Pfizer, included a pilot exploratory portion to evaluate the potential for future drug concentration monitoring using dried blood spot (DBS) sample collection. For DBS sample collection, a fixed blood volume was dispensed onto a DBS card by either regular volumetric pipette (venous blood collection) or capillary dispenser (finger prick blood collection). A 96-well semi-automated liquid-liquid extraction sample preparation procedure was developed to provide clean extracts for UHPLC-MS/MS quantitation. Assays using both partial-spot center punch and whole spot punch were developed and validated. The linear dynamic ranges for all the analyses were from 0.5 to 500 ng/mL. The coefficient of determination (r(2)) values was >0.9944 for all the validation runs. For the center punch approach, the intra-assay precision (%CV) was within 4.4% and inter-assay precision was within 2.6%. The assay accuracy, expressed as %Dev., was within ± 5.4% of the nominal concentrations. One accuracy and precision run was performed using the whole spot approach, the intra-assay precision (%CV) was within 7.1% and the accuracy was within ± 8.0% of the nominal concentrations. In contrast to the center punch approach, the whole spot approach eliminated the effect of hematocrit and high lipids on the analysis of apixaban in human DBS when an accurate sample blood volume was collected on DBS cards.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Supelco
Ethylacetat, analytical standard
Supelco
Hexan, analytical standard
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Supelco
Ethylacetat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dibutylether, anhydrous, 99.3%
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Ethylacetat
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethylacetat, ReagentPlus®, ≥99.8%