Merck
  • Development of a multi-toxin method for investigating the carryover of zearalenone, deoxynivalenol and their metabolites into milk of dairy cows.
Development of a multi-toxin method for investigating the carryover of zearalenone, deoxynivalenol and their metabolites into milk of dairy cows.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2015-04-08)
Janine Winkler, Susanne Kersten, Hana Valenta, Ulrich Meyer, Ulrich H Engelhardt, Sven Dänicke
ZUSAMMENFASSUNG
A dose-response study was carried out to examine the carryover of zearalenone (ZEN), deoxynivalenol (DON) and their metabolites into bovine milk. Therefore, a feeding trial with 30 dairy cows fed with three different levels of Fusarium (FUS) toxin-contaminated maize was performed. A control group (0.02 mg ZEN kg(-1) dry matter (DM) and 0.07 DON kg(-1) DM) was compared with two groups fed contaminated diets. The first diet contained 0.33 mg ZEN kg(-1) DM and 2.62 mg DON kg(-1) DM (group FUS-50) and the second diet contained 0.66 mg ZEN kg(-1) DM and 5.24 mg DON kg(-1) DM (group FUS-100). For milk sample analysis, a new cost-efficient sample preparation method was developed for the simultaneous determination of ZEN, DON and their metabolites. The method comprised the separation of the milk fat followed by an SPE clean-up on Oasis HLB and a LC-MS/MS measurement. The less toxic metabolite de-epoxy-DON had the highest detected concentration (5.6 ng ml(-1) milk) in the milk samples obtained from the feeding trial. Additionally, ZEN (up to 0.29 ng ml(-1)), α-zearalenol (up to 0.17 ng ml(-1)), β-zearalenol (up to 0.95 ng ml(-1)) and DON (up to 2.5 ng ml(-1)) were detected in these samples. The milk toxin concentrations of cows fed the control diet were significantly lower compared with cows fed the contaminated diet. The calculated carryover rates ranged between 0 and 0.0075 for ZEN and metabolites and between 0 and 0.0017 for DON independent of exposure. It can be concluded that dietary toxin concentrations in the feed below or close to the current guidance values do not pose a risk for consumers due to negligible carryover rates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Zirkonium(IV)-oxychlorid Octahydrat, reagent grade, 98%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%