Merck
RGD-fatty alcohol-modified docetaxel liposomes improve tumor selectivity in vivo.
International journal of pharmaceutics (2014-04-09)
Yinghuan Li, Xuelian Zheng, Yi Sun, Zhao Ren, Xuemei Li, Guohui Cui
ZUSAMMENFASSUNG
The tripeptide arginine-glycine-aspartate (RGD) was conjugated with various fatty alcohols to obtain RGDOCnH2n+1 (n=8, 10, 12, 14, 16, 18), which were incorporated into the bilayer of docetaxel liposomes to improve their tumor specificity. The fatty alcohols were accepted as linking groups to insert the tetrapeptide RGDX (X=amino acid) into the bilayer of liposomes. RGDX was previously shown to be a potent ligand to target tumor cell-surface integrin receptors, whereas RGD was not shown to have this ability. We hypothesized that RGD-fatty alcohol conjugates lacking the fourth amine X can guide liposomes to tumors without reducing their binding affinity to integrins. Antitumor activity, pharmacokinetics and biodistribution studies were evaluated in mice inoculated with S180 sarcoma. Compared with unmodified liposomes, RGD-fatty alcohol-modified liposomes successfully delivered significantly more docetaxel to tumors, which led to significant tumor weight loss and increased tumor docetaxel concentrations accompanied by reduced liver accumulation. Improved affinity of RGD-fatty alcohols to integrins was also confirmed on A375 cell model. Further comparisons among the tumor-targeting capacities of liposomes containing RGD-fatty alcohols, RGDF-fatty alcohols and RGDV-fatty acids demonstrated that RGD-fatty alcohols were as effective as the other two tetrapeptide derivatives. Therefore, a simplified tumor-targeting delivery system using RGD-fatty alcohols was developed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid, 99%
Sigma-Aldrich
4-Methylmorpholin, ReagentPlus®, 99%
Sigma-Aldrich
4-Methylmorpholin, purified by redistillation, ≥99.5%
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid -Lösung, 1.0 M in methylene chloride
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid, puriss., ≥99.0% (GC)
Sigma-Aldrich
4-Methylmorpholin, purum, ≥98.0% (GC)
Sigma-Aldrich
2-Phenylindol, technical grade, 95%
Sigma-Aldrich
4-Methylmorpholin, BioXtra, suitable for protein sequencing, ≥99.5% (GC)
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
USP
Docetaxel, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material
Wasserfreies Docetaxel, European Pharmacopoeia (EP) Reference Standard