Merck
Metabolic fate of neutral human milk oligosaccharides in exclusively breast-fed infants.
Molecular nutrition & food research (2014-10-21)
Viktoria Dotz, Silvia Rudloff, Christina Meyer, Günter Lochnit, Clemens Kunz
ZUSAMMENFASSUNG
Various biological effects have been postulated for human milk oligosaccharides (HMO), as deduced from in vitro, animal, and epidemiological studies. Little is known about their metabolic fate in vivo in the breast-fed infant, which is presented here. Human milk and infant urine and feces were collected from ten mother-child pairs and analyzed by MALDI-TOF MS (/MS), accompanied by high-performance anion-exchange chromatography with pulsed amperometric detection. Previously, we detected intact small and complex HMO in infant urine, which had been absorbed from gut, as verified via intrinsic (13) C-labeling. Our current work reveals the presence of novel HMO metabolites in urine and feces of breast-fed infants. The novel metabolites were identified as acetylated HMOs and other HMO-like structures, produced by the infants or by their gut microbiota. The finding of secretor- or Lewis-specific HMO in the feces/urine of infants fed with nonsecretor or Lewis-negative milk suggested a correspondent modification in the infant. Our study reveals new insights into the metabolism of neutral HMO in exclusively breast-fed infants and provides further indications for multiple factors influencing HMO metabolism and functions that should be considered in future in vivo investigations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Natriumacetat, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Ammoniumbicarbonat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumacetat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Natriumacetat, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Ammoniumbicarbonat, BioUltra, ≥99.5% (T)
Sigma-Aldrich
2,5-Dihydroxybenzoesäure, 98%
Sigma-Aldrich
Kreatinin, anhydrous, ≥98%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Supelco
Natronlauge, 49-51% in water, eluent for IC