Merck
  • Simultaneous determination of six flavonoids from Paulownia tomentosa flower extract in rat plasma by LC-MS/MS and its application to a pharmacokinetic study.
Simultaneous determination of six flavonoids from Paulownia tomentosa flower extract in rat plasma by LC-MS/MS and its application to a pharmacokinetic study.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-23)
Bin Dai, Zhiqiang Hu, Haiyan Li, Chong Yan, Liwei Zhang
ZUSAMMENFASSUNG
A simple, rapid and sensitive liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed and validated for simultaneous determination of six components including apigenin, quercetin, apigenin-7-O-β-D-glucoside, quercetin-3-O-β-D-glucoside, 3'-methoxyluteolin-7-O-β-D-glucoside, and tricin-7-O-β-D-glucopyranoside in rat plasma using formononetin as the internal standard (IS). The plasma samples were pretreated by a one-step liquid-liquid extraction with dichloromethane. The chromatographic separation was carried out on a ZORBAX SB-Aq column with a gradient mobile phase consisting of acetonitrile and 2mM aqueous ammonium acetate. All analytes and IS were quantitated through electrospray ionization in negative ion multiple reaction monitoring mode. The mass transitions were as follows: m/z 269.1→117.2 for apigenin, m/z 301.2→151.2 for quercetin, m/z 431.3→311.2 for apigenin-7-O-β-D-glucoside, m/z 463.2→300.2 for quercetin-3-O-β-D-glucoside, m/z 461.3→283.1 for 3'-methoxyluteolin-7-O-β-D-glucoside, m/z 491.3→313.1 for tricin-7-O-β-D-glucopyranoside, and m/z 267.2→252.2 for IS, respectively. All calibration curves exhibited good linearity with correlation coefficient (r)>0.995. The intra-day and inter-day precisions (RSD) at three QC levels were both less than 14.0% and the accuracies ranged from 89.8% to 113.8%. The extraction recoveries of six compounds ranged from 82.3% to 92.5%. The validated method was successfully applied to pharmacokinetic study of the six components in male rat plasma after oral administration of Paulownia tomentosa flower extract.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.