Merck
  • TERT promoter mutations and TERT mRNA but not FGFR3 mutations are urinary biomarkers in Han Chinese patients with urothelial bladder cancer.
TERT promoter mutations and TERT mRNA but not FGFR3 mutations are urinary biomarkers in Han Chinese patients with urothelial bladder cancer.
The oncologist (2015-02-07)
Kun Wang, Tiantian Liu, Cheng Liu, Yan Meng, Xiaotian Yuan, Li Liu, Nan Ge, Jikai Liu, Chang Wang, Hongbo Ren, Keqiang Yan, Sanyuan Hu, Zhonghua Xu, Yidong Fan, Dawei Xu
ZUSAMMENFASSUNG
The TERT promoter and FGFR3 gene mutations are two of the most common genetic events in urothelial bladder cancer (UBC), and these mutation assays in patient urine have been shown to be promising biomarkers for UBC diagnosis and surveillance. These results were obtained mainly from studies of patients with UBC in Western countries, and little is known about such information in Han Chinese patients with UBC. In the present study, we addressed this issue by analyzing tumors from 182 Han Chinese patients with UBC and urine samples from 102 patients for mutations in the TERT promoter and FGFR3 and TERT mRNA expression in tumors and/or urine. TERT promoter and FGFR3 mutations were identified in 87 of 182 (47.8%) and 7 of 102 (6.7%) UBC cases, respectively. In 46 urine samples from patients with TERT promoter mutation-carrying tumors, the mutant promoter was detected in 24 (52%) prior to operation and disappeared in most examined urine samples (80%) taken 1 week after operation. TERT mRNA was detected in urine derived from 46 of 49 patients (94%) that was analyzed before operation independently of the presence of TERT promoter mutations. Collectively, FGFR3 mutations occur at a very low rate in Han Chinese UBC and cannot serve as diagnostic markers for Chinese patients. Han Chinese patients with UBC have relatively low TERT promoter mutation frequency compared with patients in Western countries, and simultaneous detection of both mutant TERT promoter and TERT mRNA improves sensitivity and specificity of urine-based diagnosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Titan(IV)-chlorid, ReagentPlus®, 99.9% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Titan(IV)-chlorid -Lösung, 1.0 M in methylene chloride
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Titan(IV)-chlorid -Lösung, 0.09 M in 20% HCl
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Titantetrachlorid, packaged for use in deposition systems
Preise und Verfügbarkeit sind derzeit nicht verfügbar.