Merck
  • Prenatal alcohol exposure alters response of kisspeptin-ir neurons to estradiol and progesterone in adult female rats.
Prenatal alcohol exposure alters response of kisspeptin-ir neurons to estradiol and progesterone in adult female rats.
Alcoholism, clinical and experimental research (2014-11-26)
Joanna H Sliwowska, Tamara S Bodnar, Joanne Weinberg
ZUSAMMENFASSUNG
Prenatal alcohol exposure (PAE) has adverse effects on reproductive function and hypothalamic-pituitary-gonadal (HPG) activity. Kisspeptin neurons play a role in mediating feedback effects of estradiol (E2 ) and progesterone (P4 ) on the HPG axis. We hypothesized that PAE will have long-term effects on the response of kisspeptin neurons to E2 and P4 . Adult female rats (53 to 58 days) from prenatal ad libitum-fed control (C), pair-fed (PF), and alcohol-exposed (PAE) groups were subjected to Sham ovariectomy (OVX) or OVX without or with replacement with low or high physiological levels of E2 and P4 , and terminated under basal conditions. E2 and P4 levels, and the response of kisspeptin-ir neurons in the arcuate (ARC) and anteroventral periventricular (AVPV) nuclei to these hormones, were measured. As the E2 signal is conveyed to kisspeptin neurons via estrogen receptor-α (ER-α), we investigated PAE effects on the number of kisspeptin-ir/ER-α-ir neurons. To determine whether PAE alters interactions between kisspeptin and gonadotropin-releasing hormone (GnRH) neurons, close contacts between kisspeptin-ir fibers and GnRH-ir cell bodies were examined. Our data present the novel finding that kisspeptin-ir neurons in the ARC of PAE females show differential responses to E2 and to the combined treatment with E2 and P4 compared with controls: (i) OVX increased the number of kisspeptin-ir neurons in C and PF, but not PAE females compared with their Sham counterparts; (ii) E2 replacement restored kisspeptin-ir cell numbers to Sham levels in C and PF females but caused a robust down-regulation of kisspeptin-ir neurons below Sham levels in PAE females; (iii) OVX and replacement with high physiological concentrations of E2 resulted in fewer kisspeptin-ir cells in PAE than C females; (iv) OVX and replacement with high levels of both E2 and P4 markedly decreased the number of kisspeptin-ir neurons, below levels observed following E2 alone, in PF and C females, but had no significant effect in PAE females. These data suggest that a possible mechanism underlying adverse effects of PAE on HPG function involves actions of alcohol on the kisspeptin system.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
β-Estradiol, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
β-Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Wasserstoffperoxid -Lösung, tested according to Ph. Eur.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Luteinisierendes Hormon Releasing-Hormon (LHRH), ≥97% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Luteinizing hormone releasing hormone salmon, ≥97% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.