Merck
  • Separation and estimation of process-related impurities of gefitinib by reverse-phase high-performance liquid chromatography.
Separation and estimation of process-related impurities of gefitinib by reverse-phase high-performance liquid chromatography.
Journal of chromatographic science (2013-07-31)
Karunakara A Chandrashekara, Aparna Udupi, Chandrasekara G Reddy
ZUSAMMENFASSUNG
A simple and rapid reverse-phase high-performance liquid chromatographic (RP-HPLC) method was developed and validated for the simultaneous separation and estimation of gefitinib (an anti-cancer drug) and its process-related impurities. Five process-related impurities of gefitinib were separated on an Inertsil ODS-3V column (250 × 4.6 mm i.d.; particle size 5 μm) using 130 mM ammonium acetate and acetonitrile (63:37, v/v) as mobile phase (pH 5.0) with a photodiode array (PDA) detector. The correlation coefficients for gefitinib and its process-related impurities were in the range of 0.9991-0.9994 with limit of detection and limit of quantification in the range of 0.012-0.033 and 0.04-0.10 µg/mL, respectively. The recovery values were in the range of 98.26-99.90% for gefitinib and 95.99-100.55% for process-related impurities, while precision values were less than 3%. The method was found to be specific, precise and reliable for the determination of gefitinib and its impurities in gefitinib bulk drug.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammoniumacetat, ACS reagent, ≥97%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ammoniumacetat, ≥99.99% trace metals basis
Supelco
Ammoniumacetat, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Ammoniumacetat -Lösung, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammoniumacetat, 99.999% trace metals basis
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Ammoniumacetat, reagent grade, ≥98%
Sigma-Aldrich
Ammoniumacetat, BioXtra, ≥98%
Supelco
Essigsäure, analytical standard
Sigma-Aldrich
Ammoniumacetat -Lösung, BioUltra, ~5 M in H2O
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammoniumacetat, BioUltra, ≥99.0%
Supelco
Acetonitril, analytical standard