Merck
  • Total serum bilirubin predicts fat-soluble vitamin deficiency better than serum bile acids in infants with biliary atresia.
Total serum bilirubin predicts fat-soluble vitamin deficiency better than serum bile acids in infants with biliary atresia.
Journal of pediatric gastroenterology and nutrition (2014-11-25)
Veena L Venkat, Benjamin L Shneider, John C Magee, Yumirle Turmelle, Ronen Arnon, Jorge A Bezerra, Paula M Hertel, Saul J Karpen, Nanda Kerkar, Kathleen M Loomes, Jean Molleston, Karen F Murray, Vicky L Ng, Trivellore Raghunathan, Philip Rosenthal, Kathleen Schwartz, Averell H Sherker, Ronald J Sokol, Jeffrey Teckman, Kasper Wang, Peter F Whitington, James E Heubi
ZUSAMMENFASSUNG
Fat-soluble vitamin (FSV) deficiency is a well-recognized consequence of cholestatic liver disease and reduced intestinal intraluminal bile acid. We hypothesized that serum bile acid (SBA) would predict biochemical FSV deficiency better than serum total bilirubin (TB) level in infants with biliary atresia. Infants enrolled in the Trial of Corticosteroid Therapy in Infants with Biliary Atresia after hepatoportoenterostomy were the subjects of this investigation. Infants received standardized FSV supplementation and monitoring of TB, SBA, and vitamin levels at 1, 3, and 6 months. A logistic regression model was used with the binary indicator variable insufficient/sufficient as the outcome variable. Linear and nonparametric correlations were made between specific vitamin measurement levels and either TB or SBA. The degree of correlation for any particular vitamin at a specific time point was higher with TB than with SBA (higher for TB in 31 circumstances vs 3 circumstances for SBA). Receiver operating characteristic curve shows that TB performed better than SBA (area under the curve 0.998 vs 0.821). Including both TB and SBA did not perform better than TB alone (area under the curve 0.998). We found that TB was a better predictor of FSV deficiency than SBA in infants with biliary atresia. The role of SBA as a surrogate marker of FSV deficiency in other cholestatic liver diseases, such as progressive familial intrahepatic cholestasis, α-1-antitrypsin deficiency, and Alagille syndrome in which the pathophysiology is dominated by intrahepatic cholestasis, warrants further study.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Supelco
α-Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Supelco
(±)-α-Tocopherol, analytical standard
Sigma-Aldrich
Vitamin K1, viscous liquid
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Vitamin K1, BioXtra, ≥99.0% (sum of isomers, HPLC), mixture of isomers
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Ergocalciferol, ≥98.0% (sum of enantiomers, HPLC)
Supelco
Phyllochinon (K1), analytical standard
Supelco
Ergocalciferol (Vitamin D2), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Supelco
Retinol -Lösung, 100 μg/mL ± 25% (Refer to COA) (Ethanol with 0.1% (w/v) BHT), ampule of 1 mL, reference material, Cerilliant®
Supelco
Phytonadion, Pharmaceutical Secondary Standard; Certified Reference Material
USP
(±)-α-Tocopherol, United States Pharmacopeia (USP) Reference Standard
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ergocalciferol, 40,000,000 USP units/g
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Supelco
Vitamin D2 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)