Merck
  • Disruption of actin-binding domain-containing Dystonin protein causes dystonia musculorum in mice.
Disruption of actin-binding domain-containing Dystonin protein causes dystonia musculorum in mice.
The European journal of neuroscience (2014-09-10)
Masao Horie, Keisuke Watanabe, Asim K Bepari, Jun-Ichiro Nashimoto, Kimi Araki, Hiromi Sano, Satomi Chiken, Atsushi Nambu, Katsuhiko Ono, Kazuhiro Ikenaka, Akiyoshi Kakita, Ken-Ichi Yamamura, Hirohide Takebayashi
ZUSAMMENFASSUNG
The Dystonin gene (Dst) is responsible for dystonia musculorum (dt), an inherited mouse model of hereditary neuropathy accompanied by progressive motor symptoms such as dystonia and cerebellar ataxia. Dst-a isoforms, which contain actin-binding domains, are predominantly expressed in the nervous system. Although sensory neuron degeneration in the peripheral nervous system during the early postnatal stage is a well-recognised phenotype in dt, the histological characteristics and neuronal circuits in the central nervous system responsible for motor symptoms remain unclear. To analyse the causative neuronal networks and roles of Dst isoforms, we generated novel multipurpose Dst gene trap mice, in which actin-binding domain-containing isoforms are disrupted. Homozygous mice showed typical dt phenotypes with sensory degeneration and progressive motor symptoms. The gene trap allele (Dst(Gt) ) encodes a mutant Dystonin-LacZ fusion protein, which is detectable by X-gal (5-bromo-4-chloro-3-indolyl-β-D-galactoside) staining. We observed wide expression of the actin-binding domain-containing Dystonin isoforms in the central nervous system (CNS) and peripheral nervous system. This raised the possibility that not only secondary neuronal defects in the CNS subsequent to peripheral sensory degeneration but also cell-autonomous defects in the CNS contribute to the motor symptoms. Expression analysis of immediate early genes revealed decreased neuronal activity in the cerebellar-thalamo-striatal pathway in the homozygous brain, implying the involvement of this pathway in the dt phenotype. These novel Dst(Gt) mice showed that a loss-of-function mutation in the actin-binding domain-containing Dystonin isoforms led to typical dt phenotypes. Furthermore, this novel multipurpose Dst(Gt) allele offers a unique tool for analysing the causative neuronal networks involved in the dt phenotype.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Maleinsäure, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Maleinsäure, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Tetrazoliumblauchlorid, suitable for microbiology, ≥90% (T)
Supelco
Maleinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Tetrazoliumblauchlorid, used in colorimetric determination of reducing compounds
USP
Maleinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis