Merck
  • Chemical profiling and in vitro biological effects of Cardiospermum halicacabum L. (Sapindaceae) aerial parts and seeds for applications in neurodegenerative disorders.
Chemical profiling and in vitro biological effects of Cardiospermum halicacabum L. (Sapindaceae) aerial parts and seeds for applications in neurodegenerative disorders.
Journal of enzyme inhibition and medicinal chemistry (2013-10-10)
Francesco Menichini, Luigi Losi, Marco Bonesi, Alessandro Pugliese, Monica R Loizzo, Rosa Tundis
ZUSAMMENFASSUNG
Cardiospermum halicacabum is widely used in traditional medicine. Previous studies have focused on the aerial parts, while the seeds have been poorly investigated. This work aimed to analyse the chemical composition of extracts from aerial parts and seeds obtained using Naviglio and Soxhlet (PN, PS, and SN, SS, respectively), the inhibitory properties against tyrosinase, acetylcholinesterase (AChE), and butyrylcholinesterase (BChE) and the antioxidant effects. PN total extract showed significant anti-tyrosinase activity (IC50 value of 10.8 µg/mL). After partitioning with n-hexane, an HPLC method for analysing chemical constituents was established. Apigenin, luteolin, and apigenin-7-O-glucoside are the predominant constituents. SN n-hexane fraction was the most active inhibitor of BChE (IC50 of 57.9 µg/mL). Gas chromatography-mass spectrometry analysis revealed fatty acids, including eicosanoic acid, methyl 11-eicosenoate and oleic acid, as the major constituents. These findings suggest the potentiality of both seeds and aerial parts of C. halicacabum in the treatment of neurological disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Aluminiumchlorid, ReagentPlus®, 99%
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Aluminiumchlorid, reagent grade, 98%
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
p-Coumarsäure, ≥98.0% (HPLC)
Sigma-Aldrich
Kaffeesäure, ≥98.0% (HPLC)
Sigma-Aldrich
Chlorogensäure, ≥95% (titration)
Supelco
Butyliertes Hydroxytoluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butyliertes Hydroxytoluol, ≥99%, FCC, FG
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
trans-Ferulasäure, 99%
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard