Merck
Uncovering caffeine's adenosine A2A receptor inverse agonism in experimental parkinsonism.
ACS chemical biology (2014-10-01)
Víctor Fernández-Dueñas, Maricel Gómez-Soler, Marc López-Cano, Jaume J Taura, Catherine Ledent, Masahiko Watanabe, Kenneth A Jacobson, Jean-Pierre Vilardaga, Francisco Ciruela
ZUSAMMENFASSUNG
Caffeine, the most consumed psychoactive substance worldwide, may have beneficial effects on Parkinson's disease (PD) therapy. The mechanism by which caffeine contributes to its antiparkinsonian effects by acting as either an adenosine A2A receptor (A2AR) neutral antagonist or an inverse agonist is unresolved. Here we show that caffeine is an A2AR inverse agonist in cell-based functional studies and in experimental parkinsonism. Thus, we observed that caffeine triggers a distinct mode, opposite to A2AR agonist, of the receptor's activation switch leading to suppression of its spontaneous activity. These inverse agonist-related effects were also determined in the striatum of a mouse model of PD, correlating well with increased caffeine-mediated motor effects. Overall, caffeine A2AR inverse agonism may be behind some of the well-known physiological effects of this substance both in health and disease. This information might have a critical mechanistic impact for PD pharmacotherapeutic design.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
DL-Dithiothreitol, Molecular Biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Supelco
Koffein, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Koffein, powder, ReagentPlus®
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, Molecular Biology, ≥99.5% (RT)
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenosin, ≥99%
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Koffein, anhydrous, 99%, FCC, FG
Sigma-Aldrich
DL-Dithiothreitol, suitable for electrophoresis, ≥99%
Sigma-Aldrich
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
USP
Koffein, United States Pharmacopeia (USP) Reference Standard