Merck
  • Six-day selenium supplementation led to either UVA-photoprotection or toxic effects in human fibroblasts depending on the chemical form and dose of Se.
Six-day selenium supplementation led to either UVA-photoprotection or toxic effects in human fibroblasts depending on the chemical form and dose of Se.
Metallomics : integrated biometal science (2014-07-06)
Florence Hazane-Puch, Pierre Champelovier, Josiane Arnaud, Candice Trocmé, Catherine Garrel, Patrice Faure, François Laporte
ZUSAMMENFASSUNG
Selenium (Se) is an essential trace element with a narrow safety zone and unclear effects on skin photoageing. The aim of this work was to investigate the photoageing properties of sodium selenite or selenomethionine (SeMet) after a long term (6 days) Se supplementation in normal human skin fibroblasts (NHSF) subjected to ultraviolet-A (UVA) irradiation inducing 30% cell death. The uptake, toxicity and antioxidant effects of sodium selenite and SeMet were compared to better understand their photoageing properties. SeMet uptake was better than sodium selenite and their uptake by fibroblasts was not via an actively transport process. Sodium selenite induced a higher toxicity than SeMet. At 5 μM, sodium selenite inhibited cell proliferation associated with a blockage in the G2 phase and induced DNA fragmentation leading to caspase-3-dependent apoptosis cell death. At low doses (<1 μM), SeMet and sodium selenite induced glutathione peroxidase-1 (GPX1) activity and selenoproteinW1 (SEPW1) transcript expression but metalloproteinase (MMP)-1 was only induced by sodium selenite. SeMet and sodium selenite did not protect NHSFs from UVA-induced cell death. However, SeMet decreased malondialdehyde (MDA) and protected NHSFs from UVA-induced MMP1 and MMP3. We then observed a large difference in terms of photoprotection according to selenium forms. SeMet may be a potential agent for the prevention and treatment of skin photoageing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Human MMP-1 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Human MMP-1 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®