Merck
  • An ESIPT-based fluorescent probe for highly selective and ratiometric detection of mercury(II) in solution and in cells.
An ESIPT-based fluorescent probe for highly selective and ratiometric detection of mercury(II) in solution and in cells.
The Analyst (2015-02-27)
Biao Gu, Liyan Huang, Naxiu Mi, Peng Yin, Youyu Zhang, Xinman Tu, Xubiao Luo, Shenlian Luo, Shouzhuo Yao
ZUSAMMENFASSUNG
A novel ratiometric fluorescent Hg(2+) detecting system was rationally developed based on the typical excited state intramolecular proton transfer (ESIPT) characteristic of the latent fluorophore, 2-(1-(p-tolyl)-1H-phenanthro[9,10-d]imidazol-2-yl)phenol (Pol) and the Hg(2+)-mediated cleavage of the vinyl group. The probe responds selectively to Hg(2+) over various other metal ions with a larger bathochromic shift (∼100 nm). The sensing mechanism was investigated in detail by fluorescence spectroscopy, NMR spectra and mass spectrometry. Taking advantage of the enhancement effect of dichloromethane on the ESIPT efficiency, a facile dichloromethane extraction was introduced in the process of detection of Hg(2+), which affords a high sensitivity for the probe with a detection limit of 7.8 × 10(-9) M for Hg(2+). By using the new strategy, the novel probe can be used for the detection of Hg(2+) in practical water samples with good recovery. Moreover, the probe was successfully applied to the fluorescence image of Hg(2+) in living cells. These results indicated that the probe and the proposed method have promising applications for Hg(2+) sensing in biological and environmental sciences.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Kalium-tert-butoxid, reagent grade, ≥98%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Kaliumtert-butoxid -Lösung, 1.0 M in THF
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Salicylaldehyd, reagent grade, 98%
Sigma-Aldrich
1,2-Dibromethan, 98%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Supelco
Ethylacetat, analytical standard
Sigma-Aldrich
p-Toluidin, 99.6%
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
p-Toluidin, 99%
Sigma-Aldrich
1,2-Dibromethan, ≥99%
Supelco
Ethylacetat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Salicylaldehyd, redist., ≥99.0% (GC)
Sigma-Aldrich
Kalium-tert-butoxid, sublimed grade, 99.99% trace metals basis
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat