Merck
  • Microfluidic chip with thermoresponsive boronate affinity for the capture-release of cis-diol biomolecules.
Microfluidic chip with thermoresponsive boronate affinity for the capture-release of cis-diol biomolecules.
Journal of separation science (2014-11-08)
Yan Man, Guang Peng, Jianshe Wang, Xuefei Lv, Yulin Deng
ZUSAMMENFASSUNG
The polydimethylsiloxane microfluidic chip grafted with poly(N-isopropylacrylamide-3-acrylamidephenylboronic acid) (P(NIPAAm-co-AAPBA)) was fabricated by UV-induced grafting polymerization for the capture-release of cis-diol-containing biomolecules by temperature-modulated changes instead of changing the pH value of the mobile phase. Based on the optimal time for benzophenone soaking and UV irradiation of grafting polymerization, P(NIPAAm-co-AAPBA) was successfully grafted on the polydimethylsiloxane substrates, which were characterized by scanning electron microscopy, water contact angle measurements, and Fourier transform infrared spectroscopy. The P(NIPAAm-co-AAPBA)-grafted polydimethylsiloxane microfluidic chip can be successfully used for the capture and release of cis-diol-containing adenosine by adjusting the temperature from 4 to 55°C, and the result was validated by Triple Quad liquid chromatography with mass spectrometry. With further development, the fabricated polydimethylsiloxane microfluidic chips might be chosen as a potential tool for the capture and release of cis-diol-containing macromolecules, such as horseradish peroxidase and glycoprotein.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, 99%
Sigma-Aldrich
N-Isopropylacrylamid, 97%
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, ≥98.0%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, ≥98%
Sigma-Aldrich
αα-Bromisobutyrylbromid, 98%
Sigma-Aldrich
N-Isopropylacrylamid, ≥99%
Sigma-Aldrich
Poly(dimethylsiloxan), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Poly-(dimethylsiloxan), viscosity 0.65 cSt (25 °C)
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material