Merck
  • 1.1 μm superficially porous particles for liquid chromatography: part II: column packing and chromatographic performance.
1.1 μm superficially porous particles for liquid chromatography: part II: column packing and chromatographic performance.
Journal of chromatography. A (2015-01-13)
Laura E Blue, James W Jorgenson
ZUSAMMENFASSUNG
The predicted advantages of superficially porous particles over totally porous particles are decreased eddy dispersion, longitudinal diffusion, and resistance to mass transfer contributions to the theoretical plate height. While sub-2 micron superficially porous particles are commercially available, further improvements in performance are predicted by decreasing the particle diameter and decreasing the porous layer thickness. 1.1 μm superficially porous particles with 187Å pores have been synthesized using a layer-by-layer method tuned for production of smaller diameter particles. Following synthesis, these particles were packed into 30 μm i.d. capillary columns and their chromatographic performance evaluated using electrochemical detection. Based on the initial studies, the column efficiency did not meet theory, but was similar to the commercially available products tested. It is believed that the column packing process plays a critical role in the sub-par column performance. To determine if column efficiency could be predicted by solvent-particle interactions, in-solution optical microscopy and sedimentation velocity of particles in various slurry solvents were investigated and compared to column performance. Aggregating slurry solvents, such as methanol were found to produce columns with increased efficiency. The hmin for a column packed with an acetone slurry and a methanol slurry at 3mg/mL were found to be 6.3 and 3.5, respectively. Increasing the slurry concentration to 25mg/mL further improved the efficiency, producing a column with an hmin of 2.6. These efficiency results were accurately predicted by in-solution optical microscopy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Formamid, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%